missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-1,3-propanediamine, TCI America™
Chemical Identifiers
CAS |
2400-78-4 |
Molecular Formula |
C4H12N2 |
Molecular Weight (g/mol) |
88.154 |
InChI Key |
BDXGMDGYOIWKIF-UHFFFAOYSA-N |
Synonym |
1,3-Diamino-2-methylpropane |
PubChem CID |
14499052 |
IUPAC Name |
2-methylpropane-1,3-diamine |
SMILES |
CC(CN)CN |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
2400-78-4 |
|
88.154 |
|
1,3-Diamino-2-methylpropane |
|
2-methylpropane-1,3-diamine |
|
C4H12N2 |
|
BDXGMDGYOIWKIF-UHFFFAOYSA-N |
|
14499052 |
|
CC(CN)CN |
Specifications
|
Colorless |
|
2400-78-4 |
|
2734 |
|
BDXGMDGYOIWKIF-UHFFFAOYSA-N |
|
2-methylpropane-1,3-diamine |
|
14499052 |
|
Clear Liquid at 20°C |
|
2-Methyl-1,3-propanediamine |
|
1g |
|
C4H12N2 |
|
1,3-Diamino-2-methylpropane |
|
CC(CN)CN |
|
88.154 |
|
88.15 |
|
≥98.0% (GC,T) |
Safety and Handling