Please login to your online account to display your discounted pricing

2-methyl-1,3-oxazole-4-carboxylic acid, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC57901DE

 View more versions of this product

Catalog No. CC57901DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 127.1
Quantity 5g
Assay 97%
Chemical Name or Material 2-methyl-1, 3-oxazole-4-carboxylic acid
Packaging Amber Glass Bottle
Molecular Formula C5H5NO3
CAS 23062-17-1

Specifications

Chemical Name or Material 2-methyl-1, 3-oxazole-4-carboxylic acid
Assay 97%
Molecular Formula C5H5NO3
Formula Weight 127.1