missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-methyl-1,3-oxazole-4-carboxylic acid, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC57901DA
Specifications
2-methyl-1, 3-oxazole-4-carboxylic acid | |
Amber Glass Bottle | |
1g | |
IARMCEYEYXXEOS-UHFFFAOYSA-N | |
2-methyl-1,3-oxazole-4-carboxylic acid | |
539404 | |
97% |
23062-17-1 | |
C5H5NO3 | |
2-methyloxazole-4-carboxylic acid, 2-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2-methyl, 4-carboxy-2-methyl-1,3-oxazole, 2-methyloxa, oxazole-4-carboxylic acid, 2-methyl, 2-methyloxazol-4-carboxylic acid, 4-oxazolecarboxylicacid, 2-methyl, 2-methyl-3h-isoxazole-4-carboxylic acid, 2-methyl-4-oxazolecarboxylic acid | |
CC1=NC(=CO1)C(=O)O | |
127.099 | |
127.1 |
Chemical Identifiers
23062-17-1 | |
127.099 | |
2-methyloxazole-4-carboxylic acid, 2-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2-methyl, 4-carboxy-2-methyl-1,3-oxazole, 2-methyloxa, oxazole-4-carboxylic acid, 2-methyl, 2-methyloxazol-4-carboxylic acid, 4-oxazolecarboxylicacid, 2-methyl, 2-methyl-3h-isoxazole-4-carboxylic acid, 2-methyl-4-oxazolecarboxylic acid | |
2-methyl-1,3-oxazole-4-carboxylic acid |
C5H5NO3 | |
IARMCEYEYXXEOS-UHFFFAOYSA-N | |
539404 | |
CC1=NC(=CO1)C(=O)O |