Please login to your online account to display your discounted pricing

2-methyl-1,3-oxazole-4-carbaldehyde, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC57904DE

 View more versions of this product

Catalog No. CC57904DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 111.1
Quantity 5g
Assay 97%
Chemical Name or Material 2-methyl-1, 3-oxazole-4-carbaldehyde
Packaging Amber Glass Bottle
CAS 113732-84-6
Molecular Formula C5H5NO2

Specifications

Chemical Name or Material 2-methyl-1, 3-oxazole-4-carbaldehyde
Assay 97%
Molecular Formula C5H5NO2
Formula Weight 111.1