Please login to your online account to display your discounted pricing

2-Methyl-1-(3-morpholinopropyl)-5-phenyl-1H-pyrrole-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-1-(3-morpholinopropyl)-5-phenyl-1H-pyrrole-3-carboxylic acid
CAS 306936-20-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C19H24N2O3
Formula Weight 328.41
MDL Number MFCD01567236
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00344DA

 
fisher scientific company
SP00344DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.41
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-Methyl-1-(3-morpholinopropyl)-5-phenyl-1H-pyrrole-3-carboxylic acid
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C19H24N2O3
MDL Number MFCD01567236
CAS 306936-20-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.