missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2337214

 View more versions of this product

Catalog No. AAB2337214



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-1,3-dithiolane
1.15
>100°C (212°F)
C4H8S2
25 g
CARJCVDELAMAEJ-UHFFFAOYSA-N
2-methyl-1,3-dithiolane
21828
99%
5616-51-3
40°C (2 mmHg)
Unpleasant
MFCD01729666
1,3-dithiolane, 2-methyl, 2-methyl-1,3-dithiacyclopentane, unii-4o81r0spr0, 1,3-dithiolane,2-methyl, fema no. 3705, acmc-20anre, acetaldehdye ethylene thioacetal
CC1SCCS1
120.23
120.23

Chemical Identifiers

5616-51-3
120.23
CARJCVDELAMAEJ-UHFFFAOYSA-N
21828
CC1SCCS1
C4H8S2
MFCD01729666
1,3-dithiolane, 2-methyl, 2-methyl-1,3-dithiacyclopentane, unii-4o81r0spr0, 1,3-dithiolane,2-methyl, fema no. 3705, acmc-20anre, acetaldehdye ethylene thioacetal
2-methyl-1,3-dithiolane
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 227-038-2

RTECSNumber : JP1163000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only