Learn More
2-Methyl-1,3-dithiane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0020914
Description
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2-Methyl-1,3-dithiane | |
1.212 | |
82°C (179°F) | |
1.5615 | |
25 g | |
1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
2-methyl-1,3-dithiane | |
22329 | |
98% |
6007-26-7 | |
78°C to 80°C (9 mmHg) | |
C5H10S2 | |
MFCD00014652 | |
102766 | |
Not miscible or difficult to mix in water. Soluble in alcohol. | |
CC1SCCCS1 | |
134.255 | |
134.25 |
Chemical Identifiers
6007-26-7 | |
134.255 | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
22329 | |
CC1SCCCS1 |
C5H10S2 | |
MFCD00014652 | |
1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane | |
2-methyl-1,3-dithiane |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 227-859-6
RTECSNumber : JO5263000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only