missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-1,3-dithiane, 97%, ACROS Organics™
Chemical Identifiers
CAS | 6007-26-7 |
---|---|
Molecular Formula | C5H10S2 |
Molecular Weight (g/mol) | 134.26 |
InChI Key | KXROTPXCYDXGSC-UHFFFAOYSA-N |
Synonym | 1,3-dithiane, 2-methyl, 2-methyl-1,3 dithiane, 2-methyl-1,3-dithian, 2-methyl-1,3-dithiane, 2-methyl-m-dithiane, 5-19-01-00053 beilstein handbook reference, acmc-20anrf, m-dithiane, 2-methyl, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french |
PubChem CID | 22329 |
IUPAC Name | 2-methyl-1,3-dithiane |
SMILES | CC1SCCCS1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC443260010
|
Thermo Fisher Scientific
AC443260010 |
1mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC443260050
|
Thermo Fisher Scientific
AC443260050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
6007-26-7 | |
134.26 | |
1,3-dithiane, 2-methyl, 2-methyl-1,3 dithiane, 2-methyl-1,3-dithian, 2-methyl-1,3-dithiane, 2-methyl-m-dithiane, 5-19-01-00053 beilstein handbook reference, acmc-20anrf, m-dithiane, 2-methyl, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french | |
2-methyl-1,3-dithiane |
C5H10S2 | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
22329 | |
CC1SCCCS1 |
Specifications
1.1200g/mL | |
Authentic | |
1.12 | |
Colorless to Brown | |
6007-26-7 | |
100.0 | |
C5H10S2 | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
2-methyl-1,3-dithiane | |
22329 | |
Liquid | |
2-Methyl-1, 3-dithiane |
82°C | |
Glass bottle | |
66°C (5.0 Torr) | |
1mL | |
96.0 | |
96% min. (GC) | |
1,3-dithiane, 2-methyl, 2-methyl-1,3 dithiane, 2-methyl-1,3-dithian, 2-methyl-1,3-dithiane, 2-methyl-m-dithiane, 5-19-01-00053 beilstein handbook reference, acmc-20anrf, m-dithiane, 2-methyl, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french | |
CC1SCCCS1 | |
134.26 | |
134.26 | |
97% |
Safety and Handling
EINECSNumber : 227-859-6