missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-1,3-dithiane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC443260010
Specifications
2-Methyl-1, 3-dithiane | |
82°C | |
Glass bottle | |
66°C (5.0 Torr) | |
1mL | |
96.0 | |
96% min. (GC) | |
1,3-dithiane, 2-methyl, 2-methyl-1,3 dithiane, 2-methyl-1,3-dithian, 2-methyl-1,3-dithiane, 2-methyl-m-dithiane, 5-19-01-00053 beilstein handbook reference, acmc-20anrf, m-dithiane, 2-methyl, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french | |
CC1SCCCS1 | |
134.26 | |
134.26 | |
97% |
1.1200g/mL | |
Authentic | |
1.12 | |
Colorless to Brown | |
6007-26-7 | |
100.0 | |
C5H10S2 | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
2-methyl-1,3-dithiane | |
22329 | |
Liquid |
Chemical Identifiers
6007-26-7 | |
134.26 | |
1,3-dithiane, 2-methyl, 2-methyl-1,3 dithiane, 2-methyl-1,3-dithian, 2-methyl-1,3-dithiane, 2-methyl-m-dithiane, 5-19-01-00053 beilstein handbook reference, acmc-20anrf, m-dithiane, 2-methyl, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french | |
2-methyl-1,3-dithiane |
C5H10S2 | |
KXROTPXCYDXGSC-UHFFFAOYSA-N | |
22329 | |
CC1SCCCS1 |
Safety and Handling
EINECSNumber : 227-859-6