Please login to your online account to display your discounted pricing

2-Methyl-1,3-dioxolane ≥99%, Acros Organics

$477.57 - $477.57

Chemical Identifiers

Chemical Name or Material 2-Methyl-1, 3-dioxolane
Name Note 99+%
CAS 497-26-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H8O2
Formula Weight 88.11
MDL Number MFCD00003210
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC414201000

  SDS
thermo fisher scientific
414201000
Glass bottle 100g Each for $477.57
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 88.11
Physical Form Liquid
Quantity 100g
Color Clear colorless
TSCA TSCA
RTECS Number JI3509000
EINECS Number 207-841-4
Density 0.9820g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.3960 to 1.3990 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Assay Percent Range 98%
Name Note 99+%
MDL Number MFCD00003210
Molecular Formula C4H8O2
Chemical Name or Material 2-Methyl-1, 3-dioxolane
CAS 497-26-7
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.