Please login to your online account to display your discounted pricing

2-Methyl-1,3-dioxolane ≥99%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414201000

 View more versions of this product

Catalog No. AC414201000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 88.11
Quantity 100g
Color Clear colorless
TSCA TSCA
RTECS Number JI3509000
EINECS Number 207-841-4
Density 0.9820g/mL
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 2-Methyl-1, 3-dioxolane
Refractive Index 1.3960 to 1.3990 (20°C, 589nm)
Name Note 99+%
Assay Percent Range 98%
Molecular Formula C4H8O2
MDL Number MFCD00003210
CAS 497-26-7
CAS Min % 97.5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Methyl-1, 3-dioxolane
Name Note 99+%
CAS 497-26-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H8O2
Formula Weight 88.11
MDL Number MFCD00003210
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.