missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1,3-cyclopentanedione 97.0+%, TCI America™

Manufacturer:  TCI America M090125G

 View more versions of this product

Catalog No. M090125G



Specifications

Specifications

2-Methyl-1,3-cyclopentanedione
214°C
765-69-5
MFCD00001406
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methylcyclopentane-1,3-dione
13005
Crystal-Powder at 20°C
White
25g
C6H8O2
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
CC1C(=O)CCC1=O
112.128
112.13
≥97.0% (GC,T)

Chemical Identifiers

765-69-5
112.128
HXZILEQYFQYQCE-UHFFFAOYSA-N
13005
CC1C(=O)CCC1=O
C6H8O2
MFCD00001406
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
2-methylcyclopentane-1,3-dione
Safety and Handling

Safety and Handling

TSCA : No

Documents