Please login to your online account to display your discounted pricing

2-Methyl-1,3-benzothiazole-6-sulfonyl chloride, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC54703DE

 View more versions of this product

Catalog No. CC54703DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 247.73
Quantity 5g
Assay 95%
CAS 21431-13-0
Assay Percent Range 95%
Chemical Name or Material 2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
Molecular Formula C8H6ClNO2S2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6ClNO2S2
Formula Weight 247.73