Please login to your online account to display your discounted pricing

2-Methyl-1,3-benzothiazole-6-sulfonyl chloride, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6ClNO2S2
Formula Weight 247.73
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC54703DE

 
fisher scientific company
CC54703DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC54703CB

  SDS
fisher scientific company
CC54703CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC54703DA

  SDS
fisher scientific company
CC54703DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.73
Quantity 5g
Assay 95%
Chemical Name or Material 2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C8H6ClNO2S2
CAS 21431-13-0