Please login to your online account to display your discounted pricing

2-Methyl-1,3-benzothiazol-6-ylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2S
Formula Weight 164.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC54714DA

  SDS
fisher scientific company
CC54714DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC54714CB

  SDS
fisher scientific company
CC54714CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.23
Quantity 1g
Assay 97%
Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H8N2S
CAS 2941-62-0