Please login to your online account to display your discounted pricing

2-Methyl-1,3-benzothiazol-6-ylamine, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC54714CB

 View more versions of this product

Catalog No. CC54714CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250mg
Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
CAS 2941-62-0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2S
Formula Weight 164.23g/mol

Specifications

Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2S
Formula Weight 164.23g/mol