missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxytoluene, TCI America™

Manufacturer:  TCI America M0150500ML

 View more versions of this product

Catalog No. M0150500ML



Specifications

Specifications

2-Methoxytoluene
Yellow
578-58-5
MFCD00008373
2-methylanisole, o-methylanisole, 2-methoxytoluene, o-cresol methyl ether, o-cresyl methyl ether, anisole, o-methyl, benzene, 1-methoxy-2-methyl, methyl o-tolyl ether, o-methoxytoluene, 2-methylmethoxybenzene
CC1=CC=CC=C1OC
122.167
122.17
≥98.0% (GC)
171°C
500mL
C8H10O
3271
DTFKRVXLBCAIOZ-UHFFFAOYSA-N
1-methoxy-2-methylbenzene
33637
Clear Liquid at 20°C

Chemical Identifiers

578-58-5
122.167
DTFKRVXLBCAIOZ-UHFFFAOYSA-N
33637
CC1=CC=CC=C1OC
C8H10O
MFCD00008373
2-methylanisole, o-methylanisole, 2-methoxytoluene, o-cresol methyl ether, o-cresyl methyl ether, anisole, o-methyl, benzene, 1-methoxy-2-methyl, methyl o-tolyl ether, o-methoxytoluene, 2-methylmethoxybenzene
1-methoxy-2-methylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0573

TSCA : Yes

Documents