Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxythiophene, 99%

Chemical Identifiers

Chemical Name or Material 2-Methoxythiophene
CAS 16839-97-7
Molecular Formula C5H6OS
Formula Weight 114.17
MDL Number MFCD00014526
UN Number UN3271
Beilstein 106271
Synonym Methyl 2-thienyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1045506

  SDS
alfa aesar™
A1045506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1045503

  SDS
alfa aesar™
A1045503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1045514

  SDS
alfa aesar™
A1045514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.17
Assay 99%
MDL Number MFCD00014526
Molecular Formula C5H6OS
Boiling Point 154°C
Beilstein 106271
UN Number UN3271
Synonym Methyl 2-thienyl ether
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Chemical Name or Material 2-Methoxythiophene
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Health Hazard 1 Warning
Quantity 5g
Flash Point 43°C (109°F)
CAS 16839-97-7
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.