missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxythioanisole, TCI America™

$45.83 - $131.39

Chemical Identifiers

CAS 2388-73-0
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00792424
InChI Key YCUXDIFPTHNTSR-UHFFFAOYSA-N
Synonym 2-(Methylthio)anisole
PubChem CID 137577
IUPAC Name 1-methoxy-2-methylsulfanylbenzene
SMILES COC1=CC=CC=C1SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18515G View Documents TCI America
M18515G
5g Each for $45.83
 
M185125G View Documents TCI America
M185125G
25g Each for $131.39
 
Specifications

Chemical Identifiers

2388-73-0
154.227
YCUXDIFPTHNTSR-UHFFFAOYSA-N
137577
COC1=CC=CC=C1SC
C8H10OS
MFCD00792424
2-(Methylthio)anisole
1-methoxy-2-methylsulfanylbenzene

Specifications

142°C
19°C
2388-73-0
MFCD00792424
YCUXDIFPTHNTSR-UHFFFAOYSA-N
1-methoxy-2-methylsulfanylbenzene
137577
Clear Liquid at 20°C
2-Methoxythioanisole
Undesignated
5g
C8H10OS
2-(Methylthio)anisole
COC1=CC=CC=C1SC
154.227
154.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No