missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxythiazole, TCI America™

$93.20 - $279.59

Chemical Identifiers

CAS 14542-13-3
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD01631143
InChI Key MJJRDTKNLLMJDJ-UHFFFAOYSA-N
Synonym 2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
PubChem CID 575451
IUPAC Name 2-methoxy-1,3-thiazole
SMILES COC1=NC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M175325G View Documents TCI America
M175325G
25g Each for $279.59
 
M17535G View Documents TCI America
M17535G
5g Each for $93.20
 
Specifications

Chemical Identifiers

14542-13-3
115.15
MJJRDTKNLLMJDJ-UHFFFAOYSA-N
575451
COC1=NC=CS1
C4H5NOS
MFCD01631143
2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
2-methoxy-1,3-thiazole

Specifications

150°C
25g
C4H5NOS
1993
MJJRDTKNLLMJDJ-UHFFFAOYSA-N
2-methoxy-1,3-thiazole
575451
Clear Liquid at 20°C
2-Methoxythiazole
Yellow
14542-13-3
MFCD01631143
2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
COC1=NC=CS1
115.15
115.15
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No