missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxytetrahydropyran, 98%, ACROS Organics™

Chemical Identifiers

CAS 6581-66-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key XTDKZSUYCXHXJM-UHFFFAOYSA-N
Synonym 2-methoxyoxan-3-yl, 2-methoxytetrahydro-2h-pyran, 2-methoxytetrahydropyran, 2h-pyran, tetrahydro-2-methoxy, 2h-pyran,tetrahydro-2-methoxy, 5-17-03-00078 beilstein handbook reference, methoxytetrahydropyran, pyran, 2-methoxy-tetrahydro, tetrahydro-2-methoxy-2h-pyran
PubChem CID 23057
IUPAC Name 2-methoxyoxane
SMILES COC1CCCCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC450830050
SDS View Documents
Thermo Fisher Scientific
AC450830050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC450830250
SDS View Documents
Thermo Fisher Scientific
AC450830250
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6581-66-4
116.16
2-methoxyoxan-3-yl, 2-methoxytetrahydro-2h-pyran, 2-methoxytetrahydropyran, 2h-pyran, tetrahydro-2-methoxy, 2h-pyran,tetrahydro-2-methoxy, 5-17-03-00078 beilstein handbook reference, methoxytetrahydropyran, pyran, 2-methoxy-tetrahydro, tetrahydro-2-methoxy-2h-pyran
2-methoxyoxane
C6H12O2
XTDKZSUYCXHXJM-UHFFFAOYSA-N
23057
COC1CCCCO1

Specifications

0.9630g/mL
0.963
5g
97.5% min. (GC)
2-methoxyoxan-3-yl, 2-methoxytetrahydro-2h-pyran, 2-methoxytetrahydropyran, 2h-pyran, tetrahydro-2-methoxy, 2h-pyran,tetrahydro-2-methoxy, 5-17-03-00078 beilstein handbook reference, methoxytetrahydropyran, pyran, 2-methoxy-tetrahydro, tetrahydro-2-methoxy-2h-pyran
COC1CCCCO1
116.16
116.16
2-Methoxytetrahydropyran
25°C
128°C to 129°C
6581-66-4
C6H12O2
XTDKZSUYCXHXJM-UHFFFAOYSA-N
2-methoxyoxane
23057
98%
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.

GHS Signal Word: Warning

EINECSNumber : 229-509-8