missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxytetrahydropyran, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450830250
Specifications
2-Methoxytetrahydropyran | |
25°C | |
128°C to 129°C | |
6581-66-4 | |
C6H12O2 | |
XTDKZSUYCXHXJM-UHFFFAOYSA-N | |
2-methoxyoxane | |
23057 | |
98% |
0.9630g/mL | |
0.963 | |
25g | |
97.5% min. (GC) | |
2-methoxyoxan-3-yl, 2-methoxytetrahydro-2h-pyran, 2-methoxytetrahydropyran, 2h-pyran, tetrahydro-2-methoxy, 2h-pyran,tetrahydro-2-methoxy, 5-17-03-00078 beilstein handbook reference, methoxytetrahydropyran, pyran, 2-methoxy-tetrahydro, tetrahydro-2-methoxy-2h-pyran | |
COC1CCCCO1 | |
116.16 | |
116.16 |
Chemical Identifiers
6581-66-4 | |
116.16 | |
2-methoxyoxan-3-yl, 2-methoxytetrahydro-2h-pyran, 2-methoxytetrahydropyran, 2h-pyran, tetrahydro-2-methoxy, 2h-pyran,tetrahydro-2-methoxy, 5-17-03-00078 beilstein handbook reference, methoxytetrahydropyran, pyran, 2-methoxy-tetrahydro, tetrahydro-2-methoxy-2h-pyran | |
2-methoxyoxane |
C6H12O2 | |
XTDKZSUYCXHXJM-UHFFFAOYSA-N | |
23057 | |
COC1CCCCO1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS Signal Word: Warning
EINECSNumber : 229-509-8