Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxypyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Methoxypyridine
CAS 1628-89-3
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006262
UN Number UN3271
Beilstein 108189
Synonym Methyl 2-pyridyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1367514

  SDS
alfa aesar™
A1367514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1367536

  SDS
alfa aesar™
A1367536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 109.13
Beilstein 108189
Flash Point 32°C (89°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Boiling Point 142° to 143°C
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00006262
UN Number UN3271
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Quantity 25g
Health Hazard 1 Warning
Molecular Formula C6H7NO
CAS 1628-89-3
Synonym Methyl 2-pyridyl ether
Chemical Name or Material 2-Methoxypyridine
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.