Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxypyridine-5-boronic acid pinacol ester, 96%

$157.58 - $157.58

Chemical Identifiers

Chemical Name or Material 2-Methoxypyridine-5-boronic acid pinacol ester
CAS 445264-61-9
Molecular Formula C12H18BNO3
Formula Weight 235.09g/mol
MDL Number MFCD05663858
Synonym 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5013603

 
alfa aesar™
H50136-03
Each for $157.58
Description & Specifications

Specifications

Melting Point 47° to 51°C
Quantity 1g
Chemical Name or Material 2-Methoxypyridine-5-boronic acid pinacol ester
CAS 445264-61-9
Assay 96%
Molecular Formula C12H18BNO3
Formula Weight 235.09g/mol
MDL Number MFCD05663858
Synonym 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.