Learn More
Alfa Aesar™ 2-Methoxypyrazine, 98%
$20.96 - $277.58
Chemical Identifiers
CAS | 3149-28-8 |
---|---|
Molecular Formula | C5H6N2O |
Molecular Weight (g/mol) | 110.116 |
MDL Number | MFCD00006126 |
InChI Key | WKSXRWSOSLGSTN-UHFFFAOYSA-N |
Synonym | pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine |
PubChem CID | 18467 |
IUPAC Name | 2-methoxypyrazine |
SMILES | COC1=NC=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1029706 |
Alfa Aesar™
A1029706 |
5g |
Each for $65.85
|
|
AAA1029703 |
Alfa Aesar™
A1029703 |
1g |
Each for $20.96
|
|
AAA1029714 |
Alfa Aesar™
A1029714 |
25g |
Each for $277.58
|
|
Chemical Identifiers
3149-28-8 | |
110.116 | |
WKSXRWSOSLGSTN-UHFFFAOYSA-N | |
18467 | |
COC1=NC=CN=C1 |
C5H6N2O | |
MFCD00006126 | |
pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine | |
2-methoxypyrazine |
Specifications
45°C (113°F) | |
5g | |
C5H6N2O | |
UN1993 | |
pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine | |
COC1=NC=CN=C1 | |
110.116 | |
110.12 | |
2-Methoxypyrazine |
147°C to 149°C | |
3149-28-8 | |
MFCD00006126 | |
2241 | |
WKSXRWSOSLGSTN-UHFFFAOYSA-N | |
2-methoxypyrazine | |
18467 | |
98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : TSCA