Please login to your online account to display your discounted pricing

2-Methoxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxyphenylmagnesium bromide
CAS 16750-63-3
CAS Min % 18.81
CAS Max % 22.99
CAS 109-99-9
CAS Min % 77.01
CAS Max % 81.19
Molecular Formula C7H7BrMgO
Linear Formula CH3OC6H4MgBr
Formula Weight 211.34g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426738000

  SDS
thermo fisher scientific
426738000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Additional Information magnesium salts may crystallize below 20°C
Packaging AcroSeal™ Glass Bottle
Density 1.0110g/mL
Quantity 800mL
Color Clear yellow-brown
Chemical Name or Material 2-Methoxyphenylmagnesium bromide
CAS 16750-63-3
CAS Min % 18.81
CAS Max % 22.99
CAS 109-99-9
CAS Min % 77.01
CAS Max % 81.19
Physical Form Clear yellow-brown solution
Molecular Formula C7H7BrMgO
Linear Formula CH3OC6H4MgBr
Formula Weight 211.34g/mol
MDL Number MFCD01317003
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.