missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenylhydrazine Hydrochloride, TCI America™

$74.86 - $297.92

Chemical Identifiers

CAS 6971-45-5
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
MDL Number MFCD00035456
InChI Key HECSHXUNOVTAIJ-UHFFFAOYSA-N
Synonym 2-Hydrazinoanisole Hydrochloride
PubChem CID 2849445
IUPAC Name (2-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=CC=C1NN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M06175G View Documents TCI America
M06175G
5g Each for $74.86
 
M061725G View Documents TCI America
M061725G
25g Each for $297.92
 
Specifications

Chemical Identifiers

6971-45-5
174.628
HECSHXUNOVTAIJ-UHFFFAOYSA-N
2849445
COC1=CC=CC=C1NN.Cl
C7H11ClN2O
MFCD00035456
2-Hydrazinoanisole Hydrochloride
(2-methoxyphenyl)hydrazine;hydrochloride

Specifications

Undesignated
6971-45-5
MFCD00035456
HECSHXUNOVTAIJ-UHFFFAOYSA-N
(2-methoxyphenyl)hydrazine;hydrochloride
2849445
Crystal-Powder at 20°C
2-Methoxyphenylhydrazine Hydrochloride
5g
C7H11ClN2O
2-Hydrazinoanisole Hydrochloride
COC1=CC=CC=C1NN.Cl
174.628
174.63
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No