missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenylacetonitrile 98.0+%, TCI America™

Manufacturer:  TCI America M0855500G

 View more versions of this product

Catalog No. M0855500G



Specifications

Specifications

2-Methoxyphenylacetonitrile
White-Yellow
500g
C9H9NO
2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
COC1=CC=CC=C1CC#N
147.18
147.18
≥98.0% (GC)
97°C
68°C
7035-03-2
MFCD00001902
DWJKILXTMUGXOU-UHFFFAOYSA-N
2-(2-methoxyphenyl)acetonitrile
81496
Crystal-Powder at 20°C

Chemical Identifiers

7035-03-2
147.18
DWJKILXTMUGXOU-UHFFFAOYSA-N
81496
COC1=CC=CC=C1CC#N
C9H9NO
MFCD00001902
2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
2-(2-methoxyphenyl)acetonitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents