missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenylacetic Acid, TCI America™

Manufacturer:  TCI America M0533500G

 View more versions of this product

Catalog No. M0533500G



Specifications

Specifications

2-Methoxyphenylacetic Acid
124°C
93-25-4
MFCD00004321
IVEWTCACRDEAOB-UHFFFAOYSA-N
2-(2-methoxyphenyl)acetic acid
7134
Crystal-Powder at 20°C
White-Yellow
500g
C9H10O3
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
COC1=CC=CC=C1CC(=O)O
166.176
166.18
≥98.0% (T)

Chemical Identifiers

93-25-4
166.176
IVEWTCACRDEAOB-UHFFFAOYSA-N
7134
COC1=CC=CC=C1CC(=O)O
C9H10O3
MFCD00004321
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
2-(2-methoxyphenyl)acetic acid
Safety and Handling

Safety and Handling

TSCA : Yes

Documents