missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 177601000

 View more versions of this product

Catalog No. AC177601000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methoxyphenylacetic acid
98.5
122.0°C to 125.0°C
Authentic
Glass bottle
CH3OC6H4CH2CO2H
100 g
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
IVEWTCACRDEAOB-UHFFFAOYSA-N
2-(2-methoxyphenyl)acetic acid
7134
99%
93-25-4
100.0
Cream-Pink to White
99%
C9H10O3
MFCD00004321
10, 188
Solubility in water: soluble. Other solubilities: soluble in ethenol,ethyl ether,acetone
COC1=CC=CC=C1CC(O)=O
166.18
166.18
Crystalline Powder

Chemical Identifiers

93-25-4
166.18
IVEWTCACRDEAOB-UHFFFAOYSA-N
7134
COC1=CC=CC=C1CC(O)=O
C9H10O3
MFCD00004321
2-methoxyphenylacetic acid, benzeneacetic acid, 2-methoxy, 2-2-methoxyphenyl acetic acid, 2-methoxyphenylaceticacid, o-methoxyphenyl acetic acid, acetic acid, o-methoxyphenyl, 2-methoxyphenyl acetic acid, o-methoxyphenylacetic acid, acmc-209rku, 2-carboxymethyl anisole
2-(2-methoxyphenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

EINECSNumber : 202-231-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only