missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenyl Trifluoromethanesulfonate, TCI America™

$110.00 - $323.89

Chemical Identifiers

CAS 59099-58-0
Molecular Formula C8H7F3O4S
Molecular Weight (g/mol) 256.195
MDL Number MFCD25541795
InChI Key RCDZTJFBROIDKD-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2-Methoxyphenyl Ester, 2-Methoxyphenyl Triflate
PubChem CID 11780115
IUPAC Name (2-methoxyphenyl) trifluoromethanesulfonate
SMILES COC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27921G View Documents TCI America
M27921G
1g Each for $110.00
 
M27925G View Documents TCI America
M27925G
5g Each for $323.89
 
Specifications

Chemical Identifiers

59099-58-0
256.195
RCDZTJFBROIDKD-UHFFFAOYSA-N
11780115
COC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
C8H7F3O4S
MFCD25541795
Trifluoromethanesulfonic Acid 2-Methoxyphenyl Ester, 2-Methoxyphenyl Triflate
(2-methoxyphenyl) trifluoromethanesulfonate

Specifications

116°C
1g
C8H7F3O4S
Trifluoromethanesulfonic Acid 2-Methoxyphenyl Ester, 2-Methoxyphenyl Triflate
COC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
256.195
256.20
≥98.0% (GC)
Colorless
59099-58-0
MFCD25541795
RCDZTJFBROIDKD-UHFFFAOYSA-N
(2-methoxyphenyl) trifluoromethanesulfonate
11780115
Clear Liquid at 20°C
2-Methoxyphenyl Trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No