missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenyl isothiocyanate, 99%, ACROS Organics™

Chemical Identifiers

CAS 3288-04-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H7NOS
Formula Weight 165.22
MDL Number MFCD00011675
Synonym o-Anisyl isothiocyanate
Chemical Name or Material 2-Methoxyphenyl isothiocyanate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC300220050 SDS Thermo Fisher Scientific
300220050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC300220250 SDS Thermo Fisher Scientific
300220250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

3288-04-8
100.0
C8H7NOS
MFCD00011675
2-Methoxyphenyl isothiocyanate, 99%
98.5
99%
165.22
o-Anisyl isothiocyanate
≥98.5%

Specifications

3288-04-8
100.0
Authentic
Liquid
1.1900g/mL
165.22
Clear light yellow
5g
≥98.5%
98.5
Glass bottle
1.6433 to 1.6453 (20°C, 589nm)
99%
C8H7NOS
MFCD00011675
o-Anisyl isothiocyanate
2-Methoxyphenyl isothiocyanate, 99%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA