missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenyl isocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 221150050

 View more versions of this product

Catalog No. AC221150050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear colorless to slightly yellow or slightly pink
1.12g/mL
Glass bottle
1.5404 to 1.5424 (20°C, 589nm)
2-Methoxyphenyl isocyanate
700-87-8
MFCD00002004
C8H7NO2
149.15
5g
13, 378
100.0
98.5
Authentic
99%
CH3OC6H4NCO
99%

Chemical Identifiers

700-87-8
100.0
C8H7NO2
149.15
2-Methoxyphenyl isocyanate, 99%
98.5
99%
CH3OC6H4NCO
MFCD00002004
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-853-5

TSCA : TSCA