Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxyphenyl benzoate, 99%

$79.72 - $79.72

Chemical Identifiers

Chemical Name or Material 2-Methoxyphenyl benzoate
CAS 531-37-3
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD00017153
Beilstein 2053178
Synonym Benzoic acid 2-methoxyphenyl ester; Guaiacol benzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0854414

  SDS
alfa aesar™
L0854414
25g Each for $79.72
Description & Specifications

Specifications

Formula Weight 228.25
Chemical Name or Material 2-Methoxyphenyl benzoate
Assay 99%
Molecular Formula C14H12O3
MDL Number MFCD00017153
CAS 531-37-3
Synonym Benzoic acid 2-methoxyphenyl ester; Guaiacol benzoate
Beilstein 2053178
Melting Point 57° to 61°C
Quantity 25g