missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Methoxyphenoxy)acetic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442230100

 View more versions of this product

Catalog No. AC442230100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(2-Methoxyphenoxy)acetic acid
10g
97.5% min. (HPLC)
2-2-methoxyphenoxy acetic acid, 2-methoxyphenoxy acetic acid, 2-methoxyphenoxyacetic acid, acetic acid, 2-methoxyphenoxy, acetic acid, o-methoxyphenoxy, acide o-methoxyphenoxyacetique french, guaiacoxyacetic acid, o-methoxy phenoxyacetic acid, o-methoxyphenoxy acetic acid, unii-7xvz1x365a
COC1=CC=CC=C1OCC(=O)O
182.18
182.18
122°C to 123°C
1878-85-9
C9H10O4
IHONYPFTXGQWAX-UHFFFAOYSA-N
2-(2-methoxyphenoxy)acetic acid
15882
98%

Chemical Identifiers

1878-85-9
182.18
2-2-methoxyphenoxy acetic acid, 2-methoxyphenoxy acetic acid, 2-methoxyphenoxyacetic acid, acetic acid, 2-methoxyphenoxy, acetic acid, o-methoxyphenoxy, acide o-methoxyphenoxyacetique french, guaiacoxyacetic acid, o-methoxy phenoxyacetic acid, o-methoxyphenoxy acetic acid, unii-7xvz1x365a
2-(2-methoxyphenoxy)acetic acid
C9H10O4
IHONYPFTXGQWAX-UHFFFAOYSA-N
15882
COC1=CC=CC=C1OCC(=O)O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 217-526-3

SDS Documents