missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyphenethyl Alcohol, TCI America™

$55.00 - $184.86

Chemical Identifiers

CAS 7417-18-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002889
InChI Key XLDRDNQLEMMNNH-UHFFFAOYSA-N
Synonym 2-(2-Hydroxyethyl)anisole, 2-(2-Methoxyphenyl)ethanol
PubChem CID 81909
IUPAC Name 2-(2-methoxyphenyl)ethanol
SMILES COC1=CC=CC=C1CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26571G View Documents TCI America
M26571G
1g Each for $55.00
 
M26575G View Documents TCI America
M26575G
5g Each for $184.86
 
Specifications

Chemical Identifiers

7417-18-7
152.193
XLDRDNQLEMMNNH-UHFFFAOYSA-N
81909
COC1=CC=CC=C1CCO
C9H12O2
MFCD00002889
2-(2-Hydroxyethyl)anisole, 2-(2-Methoxyphenyl)ethanol
2-(2-methoxyphenyl)ethanol

Specifications

134°C
1g
C9H12O2
2-(2-Hydroxyethyl)anisole, 2-(2-Methoxyphenyl)ethanol
COC1=CC=CC=C1CCO
152.193
152.19
≥98.0% (GC)
Yellow
7417-18-7
MFCD00002889
XLDRDNQLEMMNNH-UHFFFAOYSA-N
2-(2-methoxyphenyl)ethanol
81909
Clear Liquid at 20°C
2-Methoxyphenethyl Alcohol
Safety and Handling

Safety and Handling

TSCA : No