Learn More
2-Methoxyhydroquinone, 97%, Thermo Scientific Chemicals
$93.51 - $1104.42
Chemical Identifiers
CAS | 824-46-4 |
---|---|
Molecular Formula | C7H8O3 |
Molecular Weight (g/mol) | 140.138 |
MDL Number | MFCD00013971 |
InChI Key | LAQYHRQFABOIFD-UHFFFAOYSA-N |
Synonym | 2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0 |
PubChem CID | 69988 |
IUPAC Name | 2-methoxybenzene-1,4-diol |
SMILES | COC1=C(C=CC(=C1)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1284709
|
Thermo Scientific Chemicals
A1284709 |
10 g |
Each for $93.51
|
|
|||||
AAA1284718
|
Thermo Scientific Chemicals
A1284718 |
50 g |
Each for $296.66
|
|
|||||
AAA1284730
|
Thermo Scientific Chemicals
A1284730 |
250 g |
Each for $1,104.42
|
|
|||||
Description
It finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the formation of coniferyl alcohol from ferulic acid by the white rot fungus Trametes.
Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
824-46-4 | |
140.138 | |
LAQYHRQFABOIFD-UHFFFAOYSA-N | |
69988 | |
COC1=C(C=CC(=C1)O)O |
C7H8O3 | |
MFCD00013971 | |
2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0 | |
2-methoxybenzene-1,4-diol |
Specifications
824-46-4 | |
C7H8O3 | |
10 g | |
2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0 | |
LAQYHRQFABOIFD-UHFFFAOYSA-N | |
2-methoxybenzene-1,4-diol | |
69988 | |
97% |
87°C to 91°C | |
MFCD00013971 | |
2045285 | |
Slightly soluble in water. Solubility in methanol is almost transparent. | |
COC1=C(C=CC(=C1)O)O | |
140.138 | |
140.14 | |
2-Methoxyhydroquinone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-530-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only