missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyfuran, TCI America™

Manufacturer:  TCI America M09365G

 View more versions of this product

Catalog No. M09365G



Specifications

Specifications

2-Methoxyfuran
Yellow
25414-22-6
MFCD00003227
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
COC1=CC=CO1
98.101
98.10
≥97.0% (GC)
110°C
5g
C5H6O2
1993
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
117476
Clear Liquid at 20°C

Chemical Identifiers

25414-22-6
98.101
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
117476
COC1=CC=CO1
C5H6O2
MFCD00003227
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
2-methoxyfuran
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents