missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyfuran, TCI America™

$67.22 - $279.59

Chemical Identifiers

CAS 25414-22-6
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003227
InChI Key OXCGHDNCMSOEBZ-UHFFFAOYSA-N
Synonym furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
PubChem CID 117476
IUPAC Name 2-methoxyfuran
SMILES COC1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M09361G View Documents TCI America
M09361G
1g Each for $67.22
 
M09365G View Documents TCI America
M09365G
5g Each for $279.59
 
Specifications

Chemical Identifiers

25414-22-6
98.101
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
117476
COC1=CC=CO1
C5H6O2
MFCD00003227
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
2-methoxyfuran

Specifications

110°C
1g
C5H6O2
1993
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
117476
Clear Liquid at 20°C
2-Methoxyfuran
Yellow
25414-22-6
MFCD00003227
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
COC1=CC=CO1
98.101
98.10
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator