Please login to your online account to display your discounted pricing

2-Methoxyfuran 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC160380010

 View more versions of this product

Catalog No. AC160380010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 98.1
Quantity 1g
Color Clear light yellow
EINECS Number 246-953-8
TSCA TSCA
Density 1.0600g/mL
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Refractive Index 1.4460 to 1.4510 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00003227
Molecular Formula C5H6O2
CAS 25414-22-6
Name Note 97%
Chemical Name or Material 2-Methoxyfuran
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Methoxyfuran
Name Note 97%
CAS 25414-22-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H6O2
Formula Weight 98.1
MDL Number MFCD00003227
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Highly flammable liquid and vapor.

SDS & Safety Documents