Please login to your online account to display your discounted pricing

2-Methoxyfuran 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxyfuran
Name Note 97%
CAS 25414-22-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H6O2
Formula Weight 98.1
MDL Number MFCD00003227
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160380010

  SDS
thermo fisher scientific
AC160380010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 98.1
Quantity 1g
Color Clear light yellow
EINECS Number 246-953-8
TSCA TSCA
Density 1.0600g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Methoxyfuran
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C5H6O2
MDL Number MFCD00003227
Packaging Glass bottle
CAS 25414-22-6
Refractive Index 1.4460 to 1.4510 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Highly flammable liquid and vapor.