missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxyfuran, 97%

Manufacturer:  Alfa Aesar™ A1559314

 View more versions of this product

Catalog No. AAA1559314



Specifications

Specifications

2-Methoxyfuran
110°C to 111°C
25414-22-6
MFCD00003227
106270
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
117476
97%
10°C (50°F)
25g
C5H6O2
UN3271
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
COC1=CC=CO1
98.101
98.1

Chemical Identifiers

25414-22-6
98.101
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
117476
COC1=CC=CO1
C5H6O2
MFCD00003227
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
2-methoxyfuran
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

Recommended Storage : 2° to 8°C

Documents