missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyfuran, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1559306

 View more versions of this product

Catalog No. AAA1559306



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methoxyfuran
1.065
10°C (50°F)
1.447
5 g
106270
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
117476
97%
25414-22-6
110°C to 111°C
C5H6O2
MFCD00003227
UN3271
furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
COC1=CC=CO1
98.101
98.1

Chemical Identifiers

25414-22-6
98.101
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
2-methoxyfuran
C5H6O2
MFCD00003227
117476
COC1=CC=CO1
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 246-953-8

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only