Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxyethyl phenyl sulfide, 98%

$161.62 - $161.62

Chemical Identifiers

Chemical Name or Material 2-Methoxyethyl phenyl sulfide
CAS 77298-24-9
Molecular Formula C9H12OS
Linear Formula C6H5S(CH2)2OCH3
Formula Weight 168.25
MDL Number MFCD00041329
Beilstein 5500488
Synonym 1-Methoxy-2-(phenylthio)ethane; 2-(Phenylthio)ethyl methyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1046414

  SDS
alfa aesar™
L1046414
25g Each for $161.62
Description & Specifications

Specifications

Formula Weight 168.25
Chemical Name or Material 2-Methoxyethyl phenyl sulfide
Assay 98%
Molecular Formula C9H12OS
MDL Number MFCD00041329
CAS 77298-24-9
Linear Formula C6H5S(CH2)2OCH3
Beilstein 5500488
Synonym 1-Methoxy-2-(phenylthio)ethane; 2-(Phenylthio)ethyl methyl ether
Boiling Point 125° to 126°C (18mmHg)
Quantity 25g