missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxyethyl Methanesulfonate, TCI America™

$161.95 - $467.52

Chemical Identifiers

CAS 16427-44-4
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD02656401
InChI Key BCKAHDGFNHDQST-UHFFFAOYSA-N
Synonym Methanesulfonic Acid 2-Methoxyethyl Ester
PubChem CID 146293
IUPAC Name 2-methoxyethyl methanesulfonate
SMILES COCCOS(=O)(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14465G View Documents TCI America
M14465G
5g Each for $161.95
 
M144625G View Documents TCI America
M144625G
25g Each for $467.52
 
Specifications

Chemical Identifiers

16427-44-4
154.18
BCKAHDGFNHDQST-UHFFFAOYSA-N
146293
COCCOS(=O)(=O)C
C4H10O4S
MFCD02656401
Methanesulfonic Acid 2-Methoxyethyl Ester
2-methoxyethyl methanesulfonate

Specifications

126°C
5g
C4H10O4S
Methanesulfonic Acid 2-Methoxyethyl Ester
COCCOS(=O)(=O)C
154.18
154.18
≥97.0% (GC)
Yellow
16427-44-4
MFCD02656401
BCKAHDGFNHDQST-UHFFFAOYSA-N
2-methoxyethyl methanesulfonate
146293
Clear Liquid at 20°C
2-Methoxyethyl Methanesulfonate
Safety and Handling

Safety and Handling

TSCA : No