Learn More
2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L119020B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methoxyethyl acrylate | |
3121-61-7 | |
1.008 | |
56°C (133°F) | |
C6H10O3 | |
H2C=CHCO2(CH2)2OCH3 | |
1000 g | |
1754333 | |
HFCUBKYHMMPGBY-UHFFFAOYSA-N | |
2-methoxyethyl prop-2-enoate | |
18392 | |
98% |
stabilized with ≈50 to 100ppm 4-methoxyphenol | |
<-30°C | |
162°C | |
Ester-like | |
1.428 | |
MFCD00048149,MFCD00803685 | |
UN2929 | |
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca | |
COCCOC(=O)C=C | |
130.14 | |
130.14 |
Chemical Identifiers
3121-61-7 | |
130.14 | |
HFCUBKYHMMPGBY-UHFFFAOYSA-N | |
18392 | |
COCCOC(=O)C=C |
C6H10O3 | |
MFCD00048149,MFCD00803685 | |
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca | |
2-methoxyethyl prop-2-enoate |
Safety and Handling
GHS H Statement
H311-H331-H226-H361-H302-H315-H319-H317
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P312-P330-P363-P370+P378q-P501c
H226-H302+H332-H311-H360FD
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 221-499-3
RTECSNumber : KL6000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only