Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxyethyl acetoacetate, 97%

$20.26 - $20.26

Chemical Identifiers

Chemical Name or Material 2-Methoxyethyl acetoacetate
CAS 22502-03-0
Molecular Formula C7H12O4
Formula Weight 160.17g/mol
MDL Number MFCD00009648
Beilstein 2079850
Synonym Acetoacetic acid 2-methoxyethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1062218

 
alfa aesar™
L10622-18
50g Each for $20.26
Description & Specifications

Specifications

Quantity 50g
Boiling Point 120° to 122°C (20mmHg)
Quantity 50g
Flash Point 103°C (217°F)
Chemical Name or Material 2-Methoxyethyl acetoacetate
CAS 22502-03-0
Assay 97%
Molecular Formula C7H12O4
Formula Weight 160.17g/mol
MDL Number MFCD00009648
Beilstein 2079850
Synonym Acetoacetic acid 2-methoxyethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.