Please login to your online account to display your discounted pricing

2-Methoxyethanol, ACS, 99.3+%, Alfa Aesar

$85.74 - $85.74

Chemical Identifiers

Chemical Name or Material 2-Methoxyethanol
CAS 109-86-4
Purity Grade ACS Grade
Physical Form Liquid
Assay Percent Range 99.3+%
Molecular Formula CH3OCH2CH2OH
Formula Weight 76.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA31733AK

  SDS
alfa aesar
31733AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA31733K2

  SDS
alfa aesar
31733K2
1L Each for $85.74
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 76.1
Quantity 250mL
Density 0.965
Molecular Formula CH3OCH2CH2OH
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
CAS 109-86-4
Assay Percent Range 99.3+%