Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxycrotonic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Methoxycrotonic acid
CAS 38588-37-3
Molecular Formula C5H8O3
Formula Weight 116.12
MDL Number MFCD03426027
UN Number UN3261
Beilstein 1745123
Synonym 2-Methoxy-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1929306

  SDS
alfa aesar™
L1929306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1929314

  SDS
alfa aesar™
L1929314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.12
Chemical Name or Material 2-Methoxycrotonic acid
CAS 38588-37-3
Assay 98%
UN Number UN3261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
MDL Number MFCD03426027
Beilstein 1745123
Boiling Point 117°C (16mmHg)
Melting Point 57° to 58°C
Quantity 5g
Molecular Formula C5H8O3
Synonym 2-Methoxy-2-butenoic acid
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.