missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433910050

 View more versions of this product

Catalog No. AC433910050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methoxycarbonylphenylboronic acid
Glass bottle
5g
96.0
97%
MFCD02179452
ODAXNYMENLFYMY-UHFFFAOYSA-N
(2-methoxycarbonylphenyl)boronic acid
2773496
Crystalline Powder
Authentic
White
374538-03-1
100.0
C8H9BO4
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, 2-methoxycarbonyl phenyl boronic acid, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
B(C1=CC=CC=C1C(=O)OC)(O)O
179.966
179.97
≥96.0%

Chemical Identifiers

374538-03-1
179.966
ODAXNYMENLFYMY-UHFFFAOYSA-N
2773496
B(C1=CC=CC=C1C(=O)OC)(O)O
C8H9BO4
MFCD02179452
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, 2-methoxycarbonyl phenyl boronic acid, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
(2-methoxycarbonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents